ENA308584925 Enamine

2-{4-[2-(dimethylamino)ethoxy]phenyl}ethan-1-amine

  • MDL number MFCD09047084
  • Cas Number : 100525-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69869
Availability details for
Quantity
  • Availability
  • Add to Cart