ENA308584926 Enamine

6-benzyl-2-oxo-1,2,5,6,7,8-hexahydro-1,6-naphthyridine-3-carbonitrile

  • MDL number MFCD17480464
  • Cas Number : 156365-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218...220
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69870

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69870
Availability details for
Quantity
  • Availability
  • Add to Cart