ENA308584932 Enamine

octahydro-1H-pyrido[1,2-a]piperazin-1-one

  • MDL number MFCD00910006
  • Cas Number : 15932-71-5
  • Molecular Weight : 154.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69882
Availability details for
Quantity
  • Availability
  • Add to Cart