ENA308584936 Enamine

2-oxo-1H,2H,5H,7H,8H-pyrano[4,3-b]pyridine-3-carbonitrile

  • MDL number MFCD16070469
  • Cas Number : 27152-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69889
Availability details for
Quantity
  • Availability
  • Add to Cart