ENA308584938 Enamine

5-(phenoxymethyl)thiophene-2-carboxylic acid

  • MDL number MFCD17480467
  • Cas Number : 61855-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134...136
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69891

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69891
Availability details for
Quantity
  • Availability
  • Add to Cart