ENA308584940 Enamine

1-chloro-4-(3,4-dichlorophenyl)phthalazine

  • MDL number MFCD12654796
  • Cas Number : 121004-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-69897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69897
Availability details for
Quantity
  • Availability
  • Add to Cart