ENA308584941 Enamine

1-(4-acetylthiophen-2-yl)-2-chloroethan-1-one

  • MDL number MFCD13172927
  • Cas Number : 556110-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69169

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69169
Availability details for
Quantity
  • Availability
  • Add to Cart