ENA308584943 Enamine

4-(2,2,2-trifluoroethoxy)-1,3-benzothiazol-2-amine

  • MDL number MFCD11189729
  • Cas Number : 1039884-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170...172
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69904

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69904
Availability details for
Quantity
  • Availability
  • Add to Cart