ENA308584947 Enamine

1-[(3-methylphenyl)carbonyl]piperidin-4-one

  • MDL number MFCD09938281
  • Cas Number : 1016886-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71...73
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69913

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69913
Availability details for
Quantity
  • Availability
  • Add to Cart