ENA308584954 Enamine

1,2,5-trimethylpiperidin-4-amine

  • MDL number MFCD12113779
  • Cas Number : 18103-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69930

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69930
Availability details for
Quantity
  • Availability
  • Add to Cart