ENA308584956 Enamine

1-fluoro-3-(2-isocyanoethyl)benzene

  • MDL number MFCD04117533
  • Cas Number : 730964-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69932

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69932
Availability details for
Quantity
  • Availability
  • Add to Cart