ENA308584966 Enamine

1-(1-ethyl-1H-pyrazol-4-yl)-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD12134660
  • Cas Number : 1153208-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-69962

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69962
Availability details for
Quantity
  • Availability
  • Add to Cart