ENA308584969 Enamine

potassium trifluoro(3-methoxypropyl)boranuide

  • MDL number MFCD09992908
  • Cas Number : 1445848-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69972

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-69972
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart