ENA308584976 Enamine

4-amino-5-thiatricyclo[8.4.0.0,2,6]tetradeca-1(10),2(6),3,11,13-pentaene-3-carboxamide

  • MDL number MFCD09738547
  • Cas Number : 954260-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69064

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69064
Availability details for
Quantity
  • Availability
  • Add to Cart