ENA308584980 Enamine

3-(4-butoxyphenyl)-1,2-oxazol-5-amine

  • MDL number MFCD11156298
  • Cas Number : 1021245-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-69186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69186
Availability details for
Quantity
  • Availability
  • Add to Cart