ENA308584983 Enamine

4-chloro-2,3-dihydro-1H-indazol-3-one

  • MDL number MFCD09839766
  • Cas Number : 787580-87-4
  • Molecular Weight : 168.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69187
Availability details for
Quantity
  • Availability
  • Add to Cart