ENA308584994 Enamine

tert-butyl N-[1-(5-cyanopyridin-2-yl)piperidin-4-yl]carbamate

  • MDL number MFCD17480374
  • Cas Number : 252577-86-9
  • Molecular Weight : 302.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69198

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69198
Availability details for
Quantity
  • Availability
  • Add to Cart