ENA308584997 Enamine

2-bromo-1-(4-methoxyphenyl)butan-1-one

  • MDL number MFCD00800964
  • Cas Number : 881-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69202

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69202
Availability details for
Quantity
  • Availability
  • Add to Cart