ENA308584998 Enamine

3-[(4-cyanophenyl)sulfanyl]propanoic acid

  • MDL number MFCD11135479
  • Cas Number : 381731-79-9
  • Molecular Weight : 207.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69067
Availability details for
Quantity
  • Availability
  • Add to Cart