ENA308585002 Enamine

1-(3-hydroxypropyl)-1,2-dihydropyridin-2-one

  • MDL number MFCD12186768
  • Cas Number : 25514-24-3
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69207
Availability details for
Quantity
  • Availability
  • Add to Cart