ENA308585003 Enamine

[2-(dimethylamino)-2-methylpropyl]({[4-(3-methylbutoxy)phenyl]methyl})amine

  • MDL number MFCD13351972
  • Cas Number : 1036446-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69208

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69208
Availability details for
Quantity
  • Availability
  • Add to Cart