ENA308585008 Enamine

2-(piperazin-1-yl)-N-(propan-2-yl)propanamide

  • MDL number MFCD07364122
  • Cas Number : 86906-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69211

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69211
Availability details for
Quantity
  • Availability
  • Add to Cart