ENA308585011 Enamine

4-(dimethylcarbamoyl)butanoic acid

  • MDL number MFCD11188290
  • Cas Number : 151675-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-69215

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69215
Availability details for
Quantity
  • Availability
  • Add to Cart