ENA308585015 Enamine

4-amino-N-(1-hydroxybutan-2-yl)benzamide

  • MDL number MFCD01056259
  • Cas Number : 331851-71-9
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69218

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69218
Availability details for
Quantity
  • Availability
  • Add to Cart