ENA308585026 Enamine

3-[(4-fluorophenyl)carbonyl]pyridine

  • MDL number MFCD00448413
  • Cas Number : 52779-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92...94
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69228

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69228
Availability details for
Quantity
  • Availability
  • Add to Cart