ENA308585028 Enamine

methyl 2-[4-(4-aminophenyl)piperazin-1-yl]acetate

  • MDL number MFCD11982042
  • Cas Number : 1155085-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69230

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69230
Availability details for
Quantity
  • Availability
  • Add to Cart