ENA308585029 Enamine

methyl 3-{[(3-aminophenyl)methyl]sulfanyl}propanoate

  • MDL number MFCD12092194
  • Cas Number : 1156699-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69231

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69231
Availability details for
Quantity
  • Availability
  • Add to Cart