ENA308585032 Enamine

(1S)-1-(2,5-dimethoxyphenyl)ethan-1-ol

  • MDL number MFCD09863681
  • Cas Number : 156713-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69233

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69233
Availability details for
Quantity
  • Availability
  • Add to Cart