ENA308585033 Enamine

[(3,4-difluorophenyl)(pyridin-3-yl)methyl](ethyl)amine

  • MDL number MFCD12420030
  • Cas Number : 1153288-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-69234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69234
Availability details for
Quantity
  • Availability
  • Add to Cart