ENA308585034 Enamine

[(3,4-difluorophenyl)(pyridin-3-yl)methyl](methyl)amine

  • MDL number MFCD12418677
  • Cas Number : 1153077-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-69235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69235
Availability details for
Quantity
  • Availability
  • Add to Cart