ENA308585035 Enamine

ethyl[(4-fluorophenyl)(pyridin-4-yl)methyl]amine

  • MDL number MFCD12419562
  • Cas Number : 1156223-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69236

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69236
Availability details for
Quantity
  • Availability
  • Add to Cart