ENA308585037 Enamine

4-(6-methoxypyridin-3-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD11201342
  • Cas Number : 1038337-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69237

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69237
Availability details for
Quantity
  • Availability
  • Add to Cart