ENA308585038 Enamine

[(4-fluorophenyl)(pyridin-2-yl)methyl](methyl)amine

  • MDL number MFCD12154225
  • Cas Number : 1178233-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69239

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69239
Availability details for
Quantity
  • Availability
  • Add to Cart