ENA308585061 Enamine

[(4-fluorophenyl)(pyridin-3-yl)methyl](methyl)amine

  • MDL number MFCD12181977
  • Cas Number : 381236-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69256
Availability details for
Quantity
  • Availability
  • Add to Cart