ENA308585066 Enamine

N-cyclopentylquinolin-8-amine

  • MDL number MFCD11141625
  • Cas Number : 1019549-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69262

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69262
Availability details for
Quantity
  • Availability
  • Add to Cart