ENA308585068 Enamine

4-hydroxyoxane-4-carbonitrile

  • MDL number MFCD12087857
  • Cas Number : 50289-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 39-41
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69264

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69264
Availability details for
Quantity
  • Availability
  • Add to Cart