ENA308585086 Enamine

1-ethyl-3-(3-hydroxypropyl)urea

  • MDL number MFCD00091965
  • Cas Number : 199106-80-4
  • Molecular Weight : 146.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69283

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69283
Availability details for
Quantity
  • Availability
  • Add to Cart