ENA308585087 Enamine

5-(4-bromo-3-fluorophenyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD16084789
  • Cas Number : 1250946-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69285
Availability details for
Quantity
  • Availability
  • Add to Cart