ENA308585090 Enamine

N-cyclopentyl-2-(piperazin-1-yl)propanamide

  • MDL number MFCD10011188
  • Cas Number : 1018568-87-0
  • Molecular Weight : 225.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69291

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69291
Availability details for
Quantity
  • Availability
  • Add to Cart