ENA308585092 Enamine

1-(3-aminophenyl)piperidin-4-ol

  • MDL number MFCD11101543
  • Cas Number : 1093107-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69293

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69293
Availability details for
Quantity
  • Availability
  • Add to Cart