ENA308585095 Enamine

[3-(cyclopentyloxy)-4-methoxyphenyl]methanamine

  • MDL number MFCD06213467
  • Cas Number : 151450-18-9
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69081

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69081
Availability details for
Quantity
  • Availability
  • Add to Cart