ENA308585102 Enamine

2-methyl-2-[(4-nitrophenyl)amino]propanoic acid

  • MDL number MFCD14608333
  • Cas Number : 1215813-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69297

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69297
Availability details for
Quantity
  • Availability
  • Add to Cart