ENA308585105 Enamine

[(3-amino-1,1,1-trifluoropropan-2-yl)oxy]cyclopentane

  • MDL number MFCD12047225
  • Cas Number : 1158020-65-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185...187
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69299

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-69299
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart