ENA311758939 Enamine

methyl 3-(bromomethyl)-4-fluoro-1-benzothiophene-2-carboxylate

  • MDL number MFCD18089365
  • Cas Number : 1305710-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28757

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28757
Availability details for
Quantity
  • Availability
  • Add to Cart