ENA311758942 Enamine

methyl 2-(5-amino-4H-1,2,4-triazol-3-yl)acetate

  • MDL number MFCD18089366
  • Cas Number : 875764-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-41921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41921
Availability details for
Quantity
  • Availability
  • Add to Cart