ENA311758945 Enamine

2-chloro-1-[2-(2,4-dimethoxyphenyl)pyrrolidin-1-yl]ethan-1-one

  • MDL number MFCD18089367
  • Cas Number : 1305711-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-46354

Safety Information

msds https://www.enaminestore.com/catalog/EN300-46354
Availability details for
Quantity
  • Availability
  • Add to Cart