ENA311758946 Enamine

2-chloro-1-[2-(4-ethoxyphenyl)pyrrolidin-1-yl]ethan-1-one

  • MDL number MFCD18089368
  • Cas Number : 1308646-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-46531

Safety Information

msds https://www.enaminestore.com/catalog/EN300-46531
Availability details for
Quantity
  • Availability
  • Add to Cart