ENA311758948 Enamine

2-chloro-1-[4-(4-methoxyphenyl)-1,2,3,6-tetrahydropyridin-1-yl]ethan-1-one

  • MDL number MFCD18089369
  • Cas Number : 1308650-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-46921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-46921
Availability details for
Quantity
  • Availability
  • Add to Cart