ENA311758955 Enamine

2-chloro-N-{2-[4-(trifluoromethoxy)phenyl]ethyl}acetamide

  • MDL number MFCD17976961
  • Cas Number : 1258641-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48019

Safety Information

msds https://www.enaminestore.com/catalog/EN300-48019
Availability details for
Quantity
  • Availability
  • Add to Cart