ENA311758959 Enamine

2-chloro-N-{2-[4-(trifluoromethyl)phenoxy]ethyl}acetamide

  • MDL number MFCD17976964
  • Cas Number : 1258640-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-48351

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48351
Availability details for
Quantity
  • Availability
  • Add to Cart